6-Hydroxyindazole

Lieferant: Apollo Scientific
Achtung

OR43577-1G
APOSOR43577-1GEA 27.7 EUR
APOSOR43577-1G
6-Hydroxyindazole
6-Hydroxyindazole
Formel: C₇H₆N₂O
Molecular Weight: 134,1353 g/mol
Schmelzpunkt: 217…221 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00022788
CAS-Nummer: 23244-88-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR