1-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaboranlan-2-yl)phenyl]-1H-pyrazole

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-1H-pyrazole, 3-(1H-Pyrazol-1-yl)benzeneboronic acid, pinacol ester

OR43580-250MG
APOSOR43580-250MGEA 21.9 EUR
APOSOR43580-250MG
1-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaboranlan-2-yl)phenyl]-1H-pyrazole
1-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaboranlan-2-yl)phenyl]-1H-pyrazole
Formel: C₁₅H₁₉BN₂O₂
Molecular Weight: 270,14 g/mol
Schmelzpunkt: 75…76 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07368520
CAS-Nummer: 852227-94-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR