2-(3-Methoxyphenyl)-2-methylpropanoic acid

Lieferant: Apollo Scientific
Achtung

OR43582-1G
APOSOR43582-1GEA 49.3 EUR
APOSOR43582-1G
2-(3-Methoxyphenyl)-2-methylpropanoic acid
2-(3-Methoxyphenyl)-2-methylpropanoic acid
Formel: C₁₁H₁₄O₃
Schmelzpunkt: 57…58 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09414710
CAS-Nummer: 17653-94-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR