2-(2-Methyl-1,3-thiazol-4-yl)benzoic acid

Lieferant: Apollo Scientific
Achtung

OR43586-250MG
APOSOR43586-250MGEA 116 EUR
APOSOR43586-250MG
2-(2-Methyl-1,3-thiazol-4-yl)benzoic acid
2-(2-Methyl-1,3-thiazol-4-yl)benzoic acid
Formel: C₁₁H₉NO₂S
Schmelzpunkt: 139,5…140 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09879980
CAS-Nummer: 65032-66-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR