5-Chlor-1H-indazol

Lieferant: Apollo Scientific
Achtung

OR43613-5G
APOSOR43613-5GEA 108 EUR
APOSOR43613-5G
5-Chlor-1H-indazol
5-Chloro-1H-indazole
Formel: C₇H₅ClN₂
Molecular Weight: 152,5800 g/mol
Schmelzpunkt: 118…120 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00022785
CAS-Nummer: 698-26-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR