4,5-Dimethyl-2-nitrophenol
Lieferant: Apollo Scientific
|
Achtung
|
OR43630-1G
APOSOR43630-1GEA
580
EUR
APOSOR43630-1G
4,5-Dimethyl-2-nitrophenol
4,5-Dimethyl-2-nitrophenol
Formel:
C₈H₉NO₃ Schmelzpunkt: 80…82 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD15527214 CAS-Nummer: 18087-10-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...