Glycinethylester Hydrochlorid (H-Gly-OEt.HCl) 98%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
H-Gly-OEt.HCl, Glycine Ethyl Ester HCl, Ethyl Glycinate Hydrochloride
OR43634-100G
APOSOR43634-100GEA
21.9
EUR
APOSOR43634-100G
Glycinethylester Hydrochlorid (H-Gly-OEt.HCl) 98%
Glycinethylester Hydrochlorid (H-Gly-OEt.HCl)
Formel:
C₄H₁₀ClNO₂ Molecular Weight: 139.58 g/mol Schmelzpunkt: 144…146 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012871 CAS-Nummer: 623-33-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...