5-Bromo-2-(hydroxymethyl)-1,3-thiazole 95%

Lieferant: Apollo Scientific

Synonyme: (5-Bromo-1,3-thiazol-2-yl)methanol, (5-Bromothiazol-2-yl)methanol

OR43658-250MG
APOSOR43658-250MGEA 75.4 EUR
APOSOR43658-250MG
5-Bromo-2-(hydroxymethyl)-1,3-thiazole 95%
5-Bromo-2-(hydroxymethyl)-1,3-thiazole
Formel: C₄H₄BrNOS
Molecular Weight: 194,05 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD21728921
CAS-Nummer: 911052-85-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR