3-Brom-o-toluonitril 98%

Lieferant: Apollo Scientific
Achtung

OR43660-25G
APOSOR43660-25GEA 122 EUR
APOSOR43660-25G
3-Brom-o-toluonitril 98%
3-Brom-o-toluonitril
Formel: C₈H₆BrN
Molecular Weight: 196.05 g/mol
Schmelzpunkt: 44…45 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09025665
CAS-Nummer: 52780-15-1
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR