(1H-Pyrrolo[3,2-b]pyridin-2-yl)methanol
Lieferant: Apollo Scientific
|
Achtung
|
OR43672-1G
APOSOR43672-1GEA
462
EUR
APOSOR43672-1G
(1H-Pyrrolo[3,2-b]pyridin-2-yl)methanol
(1H-Pyrrolo[3,2-b]pyridin-2-yl)methanol
Formel:
C₈H₈N₂O Schmelzpunkt: 198…200 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD18432555 CAS-Nummer: 17288-47-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...