tert-Butyl-4,5-dihydro-1H-pyrazolo[3,4-c]pyridine-6(7H)-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl 4,5-dihydro-1H-pyrazolo[3,4-c]pyridine-6(7H)-carboxylate

OR43675-250MG
APOSOR43675-250MGEA 69.6 EUR
APOSOR43675-250MG
tert-Butyl-4,5-dihydro-1H-pyrazolo[3,4-c]pyridine-6(7H)-carboxylate
tert-Butyl-4,5-dihydro-1H-pyrazolo[3,4-c]pyridine-6(7H)-carboxylate
Formel: C₁₁H₁₇N₃O₂
Molecular Weight: 223,27 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11044813
CAS-Nummer: 871726-73-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR