N-Methyl-N-(3-pyridin-3-ylbenzyl)amine 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
N-Methyl[3-(pyridin-3-yl)phenyl]methylamine
OR4368-250MG
APOSOR4368-250MGEA
203
EUR
APOSOR4368-250MG
N-Methyl-N-(3-pyridin-3-ylbenzyl)amine 97%
N-Methyl-N-(3-pyridin-3-ylbenzyl)amine
Formel:
C₁₃H₁₄N₂ Molecular Weight: 198,27 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07368550 CAS-Nummer: 852180-72-4 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...