N-Methyl-1-{4-[2-(morpholin-4-yl)ethoxy]phenyl}methylamine 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: N-Methyl-4-[2-(morpholin-4-yl)ethoxy]benzylamine, 4-(2-{4-[(Methylamino)methyl]phenoxy}ethyl)morpholine

OR4369-250MG
APOSOR4369-250MGEA 62.5 EUR
APOSOR4369-250MG
N-Methyl-1-{4-[2-(morpholin-4-yl)ethoxy]phenyl}methylamine 95%
N-Methyl-1-{4-[2-(morpholin-4-yl)ethoxy]phenyl}methylamine
Formel: C₁₄H₂₂N₂O₂
Molecular Weight: 250,337 g/mol
Siedepunkt: 141…143 °C (0,08 mm Hg)
Lagertemperatur: Raumtemperatur
MDL: MFCD07368554
CAS-Nummer: 852180-77-9
UN: 2735
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR