Methyl 3-acetyl-5,6-dihydro-4H-cyclopenta[c]thiophene-1-carboxylate 95%

Lieferant: Apollo Scientific
Achtung

OR43693-250MG
APOSOR43693-250MGEA 279 EUR
APOSOR43693-250MG
Methyl 3-acetyl-5,6-dihydro-4H-cyclopenta[c]thiophene-1-carboxylate 95%
Methyl 3-acetyl-5,6-dihydro-4H-cyclopenta[c]thiophene-1-carboxylate
Formel: C₁₁H₁₂O₃S
Molecular Weight: 224,276 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD28347696
CAS-Nummer: 1414377-89-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR