4-(3-Aminophenyl)pyridine 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(Pyridin-4-yl)aniline

OR4374-1G
APOSOR4374-1GEA 109 EUR
APOSOR4374-1G
4-(3-Aminophenyl)pyridine 97%
4-(3-Aminophenyl)pyridine
Formel: C₁₁H₁₀N₂
Molecular Weight: 170.22 g/mol
Schmelzpunkt: 165…168 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00956758
CAS-Nummer: 40034-44-4
EINECS: 254-760-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR