2-(4-Methyl-1-piperazinyl)anilin 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 1-(2-Aminophenyl)-4-methylpiperazine

OR4375-250MG
APOSOR4375-250MGEA 21.9 EUR
APOSOR4375-250MG
2-(4-Methyl-1-piperazinyl)anilin 97%
2-(4-Methyl-1-piperazinyl)aniline
Formel: C₁₁H₁₇N₃
Molecular Weight: 191,28 g/mol
Schmelzpunkt: 100,8…102.3 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04035359
CAS-Nummer: 180605-36-1
UN: 2923
ADR: 8+6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR