5-(3-Pyridinyloxy)-2-furoic acid 97%

Lieferant: Apollo Scientific
Gefahr

OR4386-1G
APOSOR4386-1GEA 53.8 EUR
APOSOR4386-1G
5-(3-Pyridinyloxy)-2-furoic acid 97%
5-(3-Pyridinyloxy)-2-furoic acid
Formel: C₁₀H₇NO₄
Molecular Weight: 205.17 g/mol
Schmelzpunkt: 150…153 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03407385
CAS-Nummer: 852180-39-3
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR