m-Tolylhydraziniumchlorid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Methylphenylhydrazin Hydrochlorid, 1-(3-Methylphenyl)hydrazin Hydrochlorid, 3-Tolylhydrazin Hydrochlorid, 3-Methylphenylhydraziniumchlorid, 1-(3-Tolyl)hydrazin Hydrochlorid, 3-Tolylhydrazinhydrochlorid, 3-Methylphenylhydrazinhydrochlorid
OR4387-5G
APOSOR4387-5GEA
29
EUR
APOSOR4387-5G
m-Tolylhydraziniumchlorid
m-Tolylhydraziniumchlorid
Formel:
C₇H₁₀N₂·HCl Molecular Weight: 158,63 g/mol Schmelzpunkt: 184…194 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012932 CAS-Nummer: 637-04-7 EINECS: 211-276-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...