Methyl-4-indolcarboxylat 98%

Lieferant: Apollo Scientific
Achtung

OR4389-1G
APOSOR4389-1GEA 21.9 EUR
APOSOR4389-1G
Methyl-4-indolcarboxylat 98%
Methyl-4-indolcarboxylat
Formel: C₁₀H₉NO₂
Molecular Weight: 175,19 g/mol
Schmelzpunkt: 68…71 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00191222
CAS-Nummer: 39830-66-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR