5-[2-(Methylthio)pyrimidin-4-yl]-thiophene-2-sulphonyl chloride

Lieferant: Apollo Scientific
Gefahr

OR4397-1G
APOSOR4397-1GEA 145 EUR
APOSOR4397-1G
5-[2-(Methylthio)pyrimidin-4-yl]-thiophene-2-sulphonyl chloride
5-[2-(Methylthio)pyrimidin-4-yl]-thiophene-2-sulphonyl chloride
Formel: C₉H₇ClN₂O₂S₃
Molecular Weight: 306,81 g/mol
Schmelzpunkt: 125…126 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00178756
CAS-Nummer: 175202-76-3
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR