1-Methylimidazol-4-carbaldehyd
Lieferant: Apollo Scientific
Synonyme:
1-Methyl-1H-imidazole-4-carbaldehyde
OR4398-1G
APOSOR4398-1GEA
470
EUR
APOSOR4398-1G
1-Methylimidazol-4-carbaldehyd
1-Methylimidazol-4-carbaldehyd
Formel:
C₅H₆N₂O Molecular Weight: 110,1139 g/mol |
MDL:
MFCD03411957 CAS-Nummer: 17289-26-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...