tert-Butyl-4-oxoazepane-1-carboxylate 95%

Lieferant: Apollo Scientific
Achtung

OR4399-1G
APOSOR4399-1GEA 21.9 EUR
APOSOR4399-1G
tert-Butyl-4-oxoazepane-1-carboxylate 95%
tert-Butyl-4-oxoazepane-1-carboxylate
Formel: C₁₁H₁₉NO₃
Lagertemperatur: Kühlschrank
MDL: MFCD03788435
CAS-Nummer: 188975-88-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR