2,4-Bis(benzyloxy)-5-bromopyrimidine 96%

Lieferant: Apollo Scientific

Synonyme: 5-Bromo-2,4-di(benzyloxy)pyrimidine

OR4413-5G
APOSOR4413-5GEA 94.4 EUR
APOSOR4413-5G
2,4-Bis(benzyloxy)-5-bromopyrimidine 96%
2,4-Bis(benzyloxy)-5-bromopyrimidine
Formel: C₁₈H₁₅BrN₂O₂
Molecular Weight: 371,2300 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00267932
CAS-Nummer: 41244-53-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR