5-Bromo-1-methyl-1H-indole
Lieferant: Apollo Scientific
|
Gefahr
|
OR4414-1G
OR4414-5G
APOSOR4414-1GEA
21.9
EUR
APOSOR4414-1G
APOSOR4414-5G
5-Bromo-1-methyl-1H-indole
5-Bromo-1-methyl-1H-indole
Formel:
C₉H₈BrN Schmelzpunkt: 39…43 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00159857 CAS-Nummer: 10075-52-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...