(1-Methyl-1H-imidazol-4-yl)methanol 97%

Lieferant: Apollo Scientific

OR4416-5G
APOSOR4416-5GEA 198 EUR
APOSOR4416-5G
(1-Methyl-1H-imidazol-4-yl)methanol 97%
(1-Methyl-1H-imidazol-4-yl)methanol
Formel: C₅H₈N₂O
Molecular Weight: 112.1298 g/mol
Schmelzpunkt: 59.5…63.5 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06738905
CAS-Nummer: 17289-25-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR