{2-Methylimidazo[1,2-a]pyridin-3-yl}methanol 90+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-(Hydroxymethyl)-2-methylimidazo[1,2-a]pyridine
OR4417-1G
APOSOR4417-1GEA
174
EUR
APOSOR4417-1G
{2-Methylimidazo[1,2-a]pyridin-3-yl}methanol 90+%
{2-Methylimidazo[1,2-a]pyridin-3-yl}methanol
Formel:
C₉H₁₀N₂O Schmelzpunkt: 157…158 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07368506 CAS-Nummer: 30489-44-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...