[5-(Thien-2-yl)-1H-pyrazol-3-yl]methanol

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-[3-(Hydroxymethyl)-1H-pyrazol-5-yl]thiophene, 3-(Hydroxymethyl)-5-(thien-2-yl)-1H-pyrazole

OR4423-1G
APOSOR4423-1GEA 108 EUR
APOSOR4423-1G
[5-(Thien-2-yl)-1H-pyrazol-3-yl]methanol
[5-(Thien-2-yl)-1H-pyrazol-3-yl]methanol
Formel: C₈H₈N₂OS
Molecular Weight: 180.227 g/mol
Schmelzpunkt: 126…128 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07368555
CAS-Nummer: 852228-02-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR