Ethyl 1-methyl-1H-indazole-3-carboxylate 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(Ethoxycarbonyl)-1-methyl-1H-indazole

OR4424-1G
APOSOR4424-1GEA 149 EUR
APOSOR4424-1G
Ethyl 1-methyl-1H-indazole-3-carboxylate 97%
Ethyl 1-methyl-1H-indazole-3-carboxylate
Formel: C₁₁H₁₂N₂O₂
Molecular Weight: 204,225 g/mol
Schmelzpunkt: 58…63 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07368499
CAS-Nummer: 220488-05-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR