4-[5-(Ethoxycarbonyl)-4-methyl-1,3-thiazol-2-yl)piperidine, N-BOC protected 90%

Lieferant: Apollo Scientific

OR4426-1G
APOSOR4426-1GEA 87 EUR
APOSOR4426-1G
4-[5-(Ethoxycarbonyl)-4-methyl-1,3-thiazol-2-yl)piperidine, N-BOC protected 90%
4-[5-(Ethoxycarbonyl)-4-methyl-1,3-thiazol-2-yl)piperidine, N-BOC protected
Formel: C₁₇H₂₆N₂O₄S
Molecular Weight: 354.464 g/mol
Schmelzpunkt: 46.6…47.1 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07368516
CAS-Nummer: 852180-50-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR