Methyl-5-(3-pyridinyloxy)-2-furoate 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR4429-1G
APOSOR4429-1GEA
97.3
EUR
APOSOR4429-1G
Methyl-5-(3-pyridinyloxy)-2-furoate 97%
Methyl-5-(3-pyridinyloxy)-2-furoate
Formel:
C₁₁H₉NO₄ Molecular Weight: 219,2 g/mol Schmelzpunkt: 78…79,5 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07368496 CAS-Nummer: 852180-38-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...