4-(3-Cyanphenyl)morpholin 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Morpholinobenzonitrile, 3-(Morpholin-4-yl)benzonitrile

OR4431-1G
APOSOR4431-1GEA 23.2 EUR
APOSOR4431-1G
4-(3-Cyanphenyl)morpholin 97%
4-(3-Cyanphenyl)morpholin
Formel: C₁₁H₁₂N₂O
Molecular Weight: 188,2300 g/mol
Schmelzpunkt: 104…106 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07368523
CAS-Nummer: 204078-31-9
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR