3-Methyl[1,2,3]triazolo[1,5-a]pyridine

Lieferant: Apollo Scientific
Achtung

OR4437-1G
APOSOR4437-1GEA 232 EUR
APOSOR4437-1G
3-Methyl[1,2,3]triazolo[1,5-a]pyridine
3-Methyl[1,2,3]triazolo[1,5-a]pyridine
Formel: C₇H₇N₃
Molecular Weight: 133,151 g/mol
Schmelzpunkt: 78…80 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08062406
CAS-Nummer: 54856-82-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR