3-Amino-5-phenyl-1H-pyrazol 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Amino-5-phenylpyrazole

OR4440-5G
APOSOR4440-5GEA 23.2 EUR
APOSOR4440-5G
3-Amino-5-phenyl-1H-pyrazol 97%
3-Amino-5-phenyl-1H-pyrazole
Formel: C₉H₉N₃
Molecular Weight: 159,19 g/mol
Schmelzpunkt: 118…121 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00191749
CAS-Nummer: 1572-10-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR