2,6-Dimethoxyphenylboronsäure
Lieferant: Apollo Scientific
|
Achtung
|
OR4443-100G
APOSOR4443-100GEA
104
EUR
APOSOR4443-100G
2,6-Dimethoxyphenylboronsäure
2,6-Dimethoxyphenylboronsäure
Formel:
C₈H₁₁BO₄ Molecular Weight: 181,98 g/mol Schmelzpunkt: 110…112 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD01318987 CAS-Nummer: 23112-96-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...