2-Amino-5-chlorpyrimidin
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
5-Chlorpyrimidin-2-ylamin
OR4453-1G
APOSOR4453-1GEA
21.9
EUR
APOSOR4453-1G
2-Amino-5-chlorpyrimidin
2-Amino-5-chloropyrimidine
Formel:
C₄H₄ClN₃ Molecular Weight: 129,55 g/mol Schmelzpunkt: 235…238 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014604 CAS-Nummer: 5428-89-7 EINECS: 226-581-2 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...