2-(Aminomethyl)piperidine, 2-BOC protected

Lieferant: Apollo Scientific
Gefahr

OR4459-250MG
APOSOR4459-250MGEA 0 EUR
APOSOR4459-250MG
2-(Aminomethyl)piperidine, 2-BOC protected
2-(Aminomethyl)piperidine, 2-BOC protected
Formel: C₁₁H₂₂N₂O₂
Schmelzpunkt: 93…97 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02181068
CAS-Nummer: 141774-61-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR