3-Bromo-2-chloropyridine-5-sulphonyl chloride 96%
Lieferant: Apollo Scientific
|
Gefahr
|
OR4475-1G
APOSOR4475-1GEA
74.1
EUR
APOSOR4475-1G
3-Bromo-2-chloropyridine-5-sulphonyl chloride 96%
3-Bromo-2-chloropyridine-5-sulphonyl chloride
Formel:
C₅H₂BrCl₂NO₂S Schmelzpunkt: 65…70 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01318107 CAS-Nummer: 216394-05-7 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...