6-Amino-5-bromopyridine-3-sulphonic acid

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-Amino-3-bromo-5-sulphopyridine

OR4477-250MG
APOSOR4477-250MGEA 21.9 EUR
APOSOR4477-250MG
6-Amino-5-bromopyridine-3-sulphonic acid
6-Amino-5-bromopyridine-3-sulphonic acid
Formel: C₅H₅BrN₂O₃S
Molecular Weight: 253,074 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03085772
CAS-Nummer: 247582-62-3
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR