(2-Acetyl-1,2-dihydroisoquinolin-1-yl)acetic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(1,2-Dihydro-2-ethanoylisoquinolin-1-yl)ethanoic acid, 2-Acetyl-1-(carboxymethyl)-1,2-dihydroisoquinoline
OR4498-100MG
APOSOR4498-100MGEA
21.9
EUR
APOSOR4498-100MG
(2-Acetyl-1,2-dihydroisoquinolin-1-yl)acetic acid
(2-Acetyl-1,2-dihydroisoquinolin-1-yl)acetic acid
Formel:
C₁₃H₁₃NO₃ Molecular Weight: 231,247 g/mol Schmelzpunkt: 163…165 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00704238 CAS-Nummer: 58246-00-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...