5-Bromopyrazolo[1,5-a]pyrimidine 97%

Lieferant: Apollo Scientific

OR450002-1G
APOSOR450002-1GEA 56.7 EUR
APOSOR450002-1G
5-Bromopyrazolo[1,5-a]pyrimidine 97%
5-Bromopyrazolo[1,5-a]pyrimidine
Formel: C₆H₄BrN₃
Molecular Weight: 198.02 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11042744
CAS-Nummer: 1159981-95-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR