2-Brom-4-methylthiazol-5-carbonsäure 95%

Lieferant: Apollo Scientific

Synonyme: 2-Brom-4-methyl-1,3-thiazol-5-carbonsäure

OR450004-25G
APOSOR450004-25GEA 102 EUR
APOSOR450004-25G
2-Brom-4-methylthiazol-5-carbonsäure 95%
2-Brom-4-methylthiazol-5-carbonsäure
Formel: C₅H₄BrNO₂S
Molecular Weight: 222.06 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09026784
CAS-Nummer: 40003-41-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR