6-Methoxy-5-nitro (1H)indazole
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Methoxy-5-nitro-1H-indazole
OR450011-1G
APOSOR450011-1GEA
149
EUR
APOSOR450011-1G
6-Methoxy-5-nitro (1H)indazole
6-Methoxy-5-nitro (1H)indazole
Formel:
C₈H₇N₃O₃ Molecular Weight: 193,159 g/mol Schmelzpunkt: 191 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11007881 CAS-Nummer: 152626-75-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...