Ethyl-4-methyl-1H-imidazol-5-carboxylat 95+%

Lieferant: Apollo Scientific

Synonyme: Ethyl 4-Methyl-1H-imidazole-5-carboxylate, 4-Methyl-1H-imidazole-5-carboxylic acid ethyl ester, Ethyl 4-methyl-5-imidazolecarboxylate, Ethyl 5-methyl-1H-imidazole-4-carboxylate

OR450016-100G
APOSOR450016-100GEA 191 EUR
APOSOR450016-100G
Ethyl-4-methyl-1H-imidazol-5-carboxylat 95+%
Ethyl-5-methyl-1H-imidazole-4-carboxylate
Formel: C₇H₁₀N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00005199
CAS-Nummer: 51605-32-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR