Ethyl ethoxyiminoacetate 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Ethyl 2-ethoxy-2-iminoacetate
OR450017-1G
OR450017-5G
OR450017-500MG
APOSOR450017-1GEA
109
EUR
APOSOR450017-1G
APOSOR450017-5G
APOSOR450017-500MG
Ethyl ethoxyiminoacetate 95%
Ethyl ethoxyiminoacetate
Formel:
C₆H₁₁NO₃ Molecular Weight: 145.16 g/mol Dichte: 1.042 g/cm³ (25 °C) Flammpunkt: 72.4 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD06797654 CAS-Nummer: 816-27-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...