Ethyl ethoxyiminoacetate 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 2-ethoxy-2-iminoacetate

OR450017-1G OR450017-5G OR450017-500MG
APOSOR450017-1GEA 109 EUR
APOSOR450017-1G APOSOR450017-5G APOSOR450017-500MG
Ethyl ethoxyiminoacetate 95%
Ethyl ethoxyiminoacetate
Formel: C₆H₁₁NO₃
Molecular Weight: 145.16 g/mol
Dichte: 1.042 g/cm³ (25 °C)
Flammpunkt: 72.4 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD06797654
CAS-Nummer: 816-27-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR