4,4,5,5-Tetramethyl-2-(tetrahydrofuran-3-yl)-1,3,2-dioxaborolane

Lieferant: Apollo Scientific
Achtung

Synonyme: Tetrahydrofuran-3-ylboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-(tetrahydro-3-furanyl)-1,3,2-Dioxaborolane, 4,4,5,5-tetramethyl-2-(oxolan-3-yl)-1,3,2-dioxaborolane

OR450023-1G
APOSOR450023-1GEA 120 EUR
APOSOR450023-1G
4,4,5,5-Tetramethyl-2-(tetrahydrofuran-3-yl)-1,3,2-dioxaborolane
4,4,5,5-Tetramethyl-2-(tetrahydrofuran-3-yl)-1,3,2-dioxaborolane
Formel: C₁₀H₁₉BO₃
Molecular Weight: 198,069 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09878900
CAS-Nummer: 331958-90-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR