tert-Butyl-5-oxo-1,4-diazepane-1-carboxylate 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Homopiperazin-5-one, N1-BOC protected, tert-Butyl 5-oxo-1,4-diazepane-1-carboxylate
OR45003-5G
APOSOR45003-5GEA
84.1
EUR
APOSOR45003-5G
tert-Butyl-5-oxo-1,4-diazepane-1-carboxylate 95%
tert-Butyl-5-oxo-1,4-diazepane-1-carboxylate
Formel:
C₁₀H₁₈N₂O₃ Molecular Weight: 214,2615 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD04115319 CAS-Nummer: 190900-21-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...