α-Brom-5-chlor-2-nitrotoluol 98%
Lieferant: Apollo Scientific
Synonyme:
2-(Bromomethyl)-4-chloro-1-nitrobenzene, (2-Bromomethyl)-4-chloro-1-nitrobenzene
OR450030-5G
APOSOR450030-5GEA
60.9
EUR
APOSOR450030-5G
α-Brom-5-chlor-2-nitrotoluol 98%
α-Brom-5-chlor-2-nitrotoluol
Formel:
C₇H₅BrClNO₂ Molecular Weight: 250,4771 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11848697 CAS-Nummer: 31577-25-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...