3,4-Dichloroisothiazole-5-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR450031-5G OR450031-25G
APOSOR450031-5GEA 65.5 EUR
APOSOR450031-5G APOSOR450031-25G
3,4-Dichloroisothiazole-5-carboxylic acid
3,4-Dichloroisothiazole-5-carboxylic acid
Formel: C₄HCl₂NO₂S
Schmelzpunkt: 175…177 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03789232
CAS-Nummer: 18480-53-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR