2-(Chloromethyl)-3,5-dimethylpyridine hydrochloride 97%

Lieferant: Apollo Scientific
Gefahr

OR450036-1G
APOSOR450036-1GEA 109 EUR
APOSOR450036-1G
2-(Chloromethyl)-3,5-dimethylpyridine hydrochloride 97%
2-(Chloromethyl)-3,5-dimethylpyridine hydrochloride
Formel: C₈H₁₀ClN·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD11112031
CAS-Nummer: 73590-93-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR